Learn More
R&D Systems™ Quantikine Immunoassay Control Set 868 for Human CCL18/PARC
Product Code. 30001963
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001963 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30001963 Supplier R&D Systems™ Supplier No. QC59B
  • ONLINE EXCLUSIVE PRICE
    2371.98 NOK
    2105.25 NOK / Pack of 1
    Save 266.73 NOK
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
For use with catalog number DCL180B, New
Applications: Control
Species Reactivity: Human
Features:
  • Quantitative controls for the determination of cytokine concentrations in biological fluids using Quantikine™ kits.
  • Contains recombinant human CCL18/PARC cytokines at low, medium and high concentrations.
  • Supplied in lyophilized form.
  • Prepared in diluted porcine serum with preservatives.
TRUSTED_SUSTAINABILITY
Spécification
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes CCL18/PARC
Sample Type Biological Fluids
Type Immunoassay Control Set
Gene ID (Entrez) 6362
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.