Learn More
R&D Systems™ Quantikine Immunoassay Control Set 571 for Human Relaxin-2
Description
Quantikine Immunoassay Control Set 571 is designed for use as a quantitative control in the determination of human Relaxin-2 concentrations in biological fluids.
- Applications: ELISA, Control
- Species Reactivity: Human
- Form: Lyophilized
- Composition: Controls are prepared in diluted porcine serum with preservatives, including recombinant human cytokines at low, medium and high concentrations.
- Storage: Store unopened product at 2°C to 8°C. Unreconstituted controls are stable for at least 6 Months from date of receipt.
- Compatibility: For use with Quantikine™ kits; specifically catalog number DRL200.
- Safety: Contains ProClin 300 (CAS No. 55965-84-9).
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Detectable Analytes | Relaxin-2 |
| Sample Type | Biological Fluids |
| Type | Immunoassay Control |
| Gene ID (Entrez) | 6019 |
| Intended Use | Control |
| Product Line | R&D Systems |
| Quantity | 1 Each |
| Source | Human |
| Form | Lyophilized |