Learn More
Thermo Scientific™ Quantech™ Fluorometer Narrow-Band Excitation and Emission Filters
Thermo Scientific™ Quantech Digital Filter Fluorometer Narrow Band Filters allow you to choose the best excitation and emission wavelength ranges for your fluorescence experiment.
3493.50 NOK - 11969.70 NOK
Specifications
Type Narrow Band Filter
For Use With (Equipment) Quantech
View More Specs
Products 17
Product Code Brand Wavelength Price Quantity & Availability  
Product Code Brand Wavelength Price Quantity & Availability  
11721111
View Documents Promotion Details
Thermo Scientific™
LE1095X1
254 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11731111
View Documents Promotion Details
Thermo Scientific™
LE1095X10
440 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11741111
View Documents Promotion Details
Thermo Scientific™
LE1095X11
450 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11751111
View Documents Promotion Details
Thermo Scientific™
LE1095X12
460 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11651359
View Documents Promotion Details
Thermo Scientific™
LE1095X13
490 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11761111
View Documents Promotion Details
Thermo Scientific™
LE1095X14
520 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11771111
View Documents Promotion Details
Thermo Scientific™
LE1095X15
540 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11781111
View Documents Promotion Details
Thermo Scientific™
LE1095X16
590 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11791111
View Documents Promotion Details
Thermo Scientific™
LE1095X17
680 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11701121
View Documents Promotion Details
Thermo Scientific™
LE1095X3
297 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11711121
View Documents Promotion Details
Thermo Scientific™
LE1095X30
360 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10324110
View Documents Promotion Details
Thermo Scientific™
LE1095X35
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10739978
View Documents Promotion Details
Thermo Scientific™
LE1095X4
313 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11771121
View Documents Promotion Details
Thermo Scientific™
LE1095X6
365 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11781121
View Documents Promotion Details
Thermo Scientific™
LE1095X7
390 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11791121
View Documents Promotion Details
Thermo Scientific™
LE1095X8
405 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11701131
View Documents Promotion Details
Thermo Scientific™
LE1095X9
420 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Thermo Scientific™ Quantech Digital Filter Fluorometer Narrow Band Filters allow you to choose the best excitation and emission wavelength ranges for your fluorescence experiment. Narrow band filters allow a narrow range of light to pass through the filter -- delivering the selectivity you need for accurate fluorescence assays.
Narrow band filters can be used to excite fluorescence or to measure a very small window of fluorescence emission. To select the proper filter for your assay, use the following guidelines:
  • There should be no overlap between the Narrow Band excitation filter and the Sharp Cut emission filter
  • A portion of the fluorescence excitation (absorption) spectrum near the peak falls in the wavelength range transmitted by the Narrow Band excitation filter
For the most sensitive measurement of fluorescence, an emission filter that does not overlap with this excitation filter must be selected.
Specifications
Specifications
Narrow Band Filter
Quantech