Learn More
Sampling Systems™ Quality Control PharmaLabels
Product Code. 15869491 Shop All Sampling Systems Products
Change view
Click to view available options
Dimensions (LxW):
95 x 95 mm
150 x 150 mm
Color:
Black
Blue
Green
Red
White
Yellow
Display Text:
Blank
Quality Control
Sampled
No. per Pack:
250/Pk.
500/Pk.
Unit Size:
1 roll
Pack of 250
Pack of 500
Product Code. Dimensions (LxW) Color Display Text No. per Pack unitSize
15869491 150 x 150 mm Red Quality Control 250/Pk. Pack of 250
15188385 150 x 150 mm White Blank 250/Pk. 1 roll
13245270 95 x 95 mm Green Quality Control 500/Pk. Pack of 500
15158395 150 x 150 mm Yellow Quality Control 250/Pk. 1 roll
15198668 95 x 95 mm Yellow Quality Control 500/Pk. 1 roll
15809501 95 x 95 mm Red Quality Control 500/Pk. Pack of 500
15819501 150 x 150 mm Blue Sampled 250/Pk. Pack of 250
15829501 95 x 95 mm Blue Quality Control 500/Pk. Pack of 500
15839501 95 x 95 mm Black Quality Control 500/Pk. Pack of 500
15849501 95 x 95 mm White Blank 500/Pk. Pack of 500
15859501 95 x 95 mm Blue Sampled 500/Pk. Pack of 500
15879491 150 x 150 mm Green Quality Control 250/Pk. Pack of 250
15889491 150 x 150 mm Blue Quality Control 250/Pk. Pack of 250
15899491 150 x 150 mm Black Quality Control 250/Pk. Pack of 250
14 options
Product Code. 15869491
Brand: Sampling Systems™ 8220P001
Only null left
Add to basket
Add to basket
Medium strength adhesion paper labels with a special FDA acceptable adhesive. Sampling Systems™ Quality Control PharmaLabels are ideal for applications where the adhesive might come into contact with the product.
  • Medium strength adhesive
  • Large writing area
  • Each roll individually packed
  • FDA acceptable adhesive (CFR 175.105)
  • Ideal for applications where the adhesive might come into contact with the product
Specifications
Certifications/Compliance CFR 175.105
Description Food Grade Adhesive
Color Red
Material Paper
Product Type PharmaLabel
Display Text Quality Control
No. per Pack 250/Pk.
Temperature Range (Metric) -40°C to +70°C
Dimensions (LxW) 150 x 150 mm