Learn More
Socorex™ Virgin Polypropylene Micropipette Tip
Description
- Natural macrotip made of virgin polypropylene
- 10mL
- Disposable
Specifications
Specifications
| Volume (Metric) | 10 mL |
| Sterility | Sterile |
| Color | Natural |
| Disposable | Single-use |
| For Use With (Equipment) | Universal Fit |
| Material | Virgin Polypropylene |