Learn More
Sartorius Stedim Biotech™ Qualitative/Technical Papers, Smooth, Discs, Grade 3 m/N
Shop All Sartorius Products
Click to view available options
Diameter (Metric):
55 mm
70 mm
90 mm
230 mm
320 mm
Unit Size:
Pack of 100
Description
- Made of refined pulp and linters
- Medium fast filtering
- Non-sterile and bulk packed
Specifications
Specifications
| Basis Weight | 65 g/m2 |
| Format | Disc |
| Material | Cotton Linters, Refined Pulp |
| Particle Retention | 7 to 10 μm |
| Filtration Speed | 30 sec. |
| Grade | 3 m/N |
| Quantity | 100/Pk. |
| Thickness (Metric) | 0.14 mm |
| Diameter (Metric) | 90 mm |