Learn More
Sartorius Qualitative Filter Paper Discs
These Qualitative Filter Papers are especially suitable for qualitative analyses, analytical purposes and routine analyses that require a low ash content.
104.00 NOK - 800.00 NOK
Specifications
Material | Cotton Linters, Refined Pulp |
---|---|
Fit for Purpose | Certified Quality |
Product Code | Brand | Diameter (Metric) | Particle Retention | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Particle Retention | Price | Quantity & Availability | ||||
15902890
|
Sartorius
FT-3-205-037 |
37 mm | 5 to 8 μm |
104.00 NOK
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15912890
|
Sartorius
FT-3-205-047 |
47 mm | 5 to 8 μm |
131.00 NOK
Pack of 100 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
15912900
|
Sartorius
FT-3-206-320 |
320 mm | 12 to 15 μm |
800.00 NOK
Pack of 100 |
Estimated Shipment: 11-07-2024
Log in to see stock. |
||||
Description
- Technical specifications:
- Made of cotton linters
- Low-nitrogen and nitrates content
- Ash content ≤0.06%
- Weight: 87 g/cm2
- Thickness: 0.18 mm
- No additives, such as wet-strengthening
- Example of applications:
- Determination of nitrogen content in soils
- Malt filtration according to EBC standards
Specifications
Cotton Linters, Refined Pulp | |
Certified Quality |