Learn More
Macherey-Nagel™ Qualitative Analysis Test Papers, MN 615
Product Code. 10402275 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
82 mm
85 mm
Unit Size:
Pack of 100
Product Code. 10402275
Brand: Macherey-Nagel™ 4310082
Only null left
Add to basket
Add to basket
Widely used for routine separations.
  • Qualitative filter papers widely used for routine separations
  • Manufactured from top quality raw materials using ultra pure water to ensure consistent and reliable filtration
  • Most widely used general purpose filter paper with medium retention and flow rate
  • This filter paper covers a wide range of laboratory applications and is frequently used for clarifying liquids
Specifications
Format Disc
Grade MN 615
Quantity 100/Pk.
Diameter (Metric) 82 mm