Learn More
Macherey-Nagel™ Qualitative Analysis Test Papers, MN 615
Product Code. 10187374 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
82 mm
85 mm
Unit Size:
Pack of 100
Product Code. Diameter (Metric) unitSize
10187374 20 mm Pack of 100
10758460 10 mm Pack of 100
10391255 15 mm Pack of 100
10499634 25 mm Pack of 100
10320415 30 mm Pack of 100
10402275 82 mm Pack of 100
10176087 85 mm Pack of 100
7 options
Product Code. 10187374
Brand: Macherey-Nagel™ 431002
Only null left
Add to basket
Add to basket
Widely used for routine separations.
  • Qualitative filter papers widely used for routine separations
  • Manufactured from top quality raw materials using ultra pure water to ensure consistent and reliable filtration
  • Most widely used general purpose filter paper with medium retention and flow rate
  • This filter paper covers a wide range of laboratory applications and is frequently used for clarifying liquids
Specifications
Format Disc
Grade MN 615
Quantity 100/Pk.
Diameter (Metric) 20 mm