Learn More
Qsonica Q700/Q500 Accessories: 8 Tube Holder Rack
Product Code. p-4912121 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
8 x 1.5 mL Polypropylene Eppendorf Tubes
Color:
Green
Unit Size:
Each
Product Code. Holds Color unitSize
15390372 8 x 1.5 mL Polypropylene Eppendorf Tubes Green Each
1 options
Product Code. 15390372
Only null left
Add to basket
Add to basket
An excellent solution for securely holding tubes in place when working with Q500/Q700 Sonicators.
  • Holds 8 tubes of 1.5mL
  • Available for either Evergreen or Eppendorf tubes
Specifications
Product Type Tube Holder Rack
Color Green
For Use With (Equipment) Cup Horn (Part # 431C2)
Holds 8 x 1.5 mL Polypropylene Eppendorf Tubes