Learn More
Hellma™ Suprasil™ Quartz Micro Absorption Cell Cuvettes
Product Code. 13227019
Change view
Click to view available options
Capacity (Metric):
14,000 μL
17,500 μL
7000 μL
Unit Size:
Each
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
13227019 17,500 μL Each
10028190 14,000 μL Each
10100591 7000 μL Each
11757263 7000 μL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 13227019 Supplier Hellma™ Supplier No. 1005040
  • ONLINE EXCLUSIVE PRICE
    2875.00 NOK
    2659.33 NOK / Pack of 1
    Save 215.67 NOK
    7% Off
Estimated Shipment: 21-12-2026
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Suprasil™ Quartz Micro Absorption Cell Cuvettes come with either a lid or PTFE cap to limit evaporation.
  • Very chemically resistant
  • Maintains shape, even at high temperatures up to approximately 1000°C
  • 80% transmission from 320 to 2500nm
Specifications
Type Macro Absorption Cell
Capacity (Metric) 17,500 μL
Material Quartz Suprasil
Width (Metric) Interior 9.5mm
Path Length 50 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 45 mm
Width (Metric) 12.5 mm