Learn More
Hellma™ QS Quartz Suprasil High Precision Macro Absorption Cells With PTFE Stopper
Product Code. 11747273
Click to view available options
Capacity (Metric):
350 μL
7000 μL
Unit Size:
Each
Pack of 2
Product Code. 11747273
Brand: Hellma™ 1102040
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Macro Absorption Cell
Capacity (Metric) 7000 μL
Material Quartz Suprasil
Closure Material PTFE
Width (Metric) Interior 9.5mm
Path Length 20 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 46 mm
Width (Metric) 12.5 mm
For Use With (Application) Photometric and Colorimetric Measurements