Learn More
MP Biomedicals™ qPCR & Go SYBR Hi-ROX Kit
Shop All MP Biomedicals Products
Click to view available options
No. of Reactions:
2000 Reactions
500 Reactions
Unit Size:
2000 reactions
500 reactions
Description
- The qPCR & Go SYBR™ Hi-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
- A combination of the latest advances in buffer chemistry and enhancers, together with an antibody-mediated hot-start DNA polymerase system, ensures that the qPCR & Go SYBR™ Hi-ROX Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
- For ease-of-use and added convenience, qPCR & Go SYBR™ Hi-ROX is provided as a 2x mastermix containing all the components necessary for real-time PCR, including the SYBR™Green I dye, dNTPs, stabilisers and enhancers.
- The kit consists of a ready-to-use premix, only primers and template need to be added.
Specifications
Specifications
| Content And Storage | -20°C |
| Format | Kit |
| Product Type | qPCR Kit |
| Quantity | 5 x 1 mL |
| For Use With (Application) | Real Time PCR (qPCR) |
| No. of Reactions | 500 Reactions |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.