Learn More
asecos™ Q-PHOENIX-90 Yellow Safety Storage Cabinets
Product Code. 15937416 Shop All asecos Products
Click to view available options
Configuration:
3 Shelves, 1 Insert, 1 Bottom Collecting Sump
3 Tray Shelves, 1 Perforated Insert (365 mm), 1 Bottom Collecting Sump
4 Drawers
6 Drawers
Unit Size:
Each
Product Code. 15937416
Brand: asecos™ 3214904033647
This item requires a price update prior to being ordered.
Request a Price for this item here.
For the safe and approved storage of flammable hazardous substances in working areas with fire resistance up to 90 minutes.
  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Space saving folding doors: less required space in front of the cabinet while doors are open
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Specifications
Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 4 Drawers
Depth (Metric) Exterior 615 mm
Depth (Metric) Interior 522 mm
Dimensions (D x W x H) Interior 522 x 1050 x 1647 mm
Dimensions (D x W x H) 615 x 1193 x 1953 mm
Door Style Folding
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 1953 mm
Height (Metric) Interior 1647 mm
Includes Entry height transport base
Model Q90.195.120.FD
No. of Doors 2
Surface Coating Powder-coated textured
Weight (Metric) 505 kg
Width (Metric) Exterior 1193 mm
Width (Metric) Interior 1050 mm
Color Anthracite Gray, Warning Yellow
Height (Metric) Opening 90 mm
Material Steel
Product Type Flammables Storage Cabinet