Learn More
Asecos™ Q-CLASSIC-90 Two-Door Light Gray Safety Storage Cabinets
Product Code. 15870448 Shop All asecos Products
Change view
Click to view available options
Load Bearing Capacity:
4 x 60 kg/19 L Drawers
4 x 60 kg/29 L Drawers
75 kg Shelves, 75 kg Insert, 33 L Sump
Unit Size:
Each
Product Code. Load Bearing Capacity unitSize
15870448 4 x 60 kg/29 L Drawers Each
15860448 75 kg Shelves, 75 kg Insert, 33 L Sump Each
15880448 4 x 60 kg/19 L Drawers Each
3 options
Product Code. 15870448
Brand: Asecos™ 3000104133647
This item requires a price update prior to being ordered.
Request a Price for this item here.
For the safe and approved storage of flammable hazardous substances in working areas.
  • Extremely robust design with scratch-proof surface
  • Triple-hinged doors
  • Safety elements assembled outside the storage compartment for increased protection against corrosion
  • Integrated transport base for simple in-house transport
  • Cabinet can be transported upright through doors
  • Adjustable feet in base allow the safe and fast compensation of minor surface unevenness
  • Modern bicolor design - choice of seven different door colors
  • Language-free labelling for optimum international usability
  • Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
  • Locking status indicator (red/green) signals the locking status
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • In accordance with EN 14470-1 and TRGS 510
  • Fire resistance of 90 minutes
Specifications
Certifications/Compliance EN 14470-1
Dimensions (D x W x H) 615 x 1193 x 1953 mm
Door Style Triple-hinged
For Use With (Application) Storage of flammable hazardous substances
Load Bearing Capacity 4 x 60 kg/29 L Drawers
No. of Doors 2
Weight (Metric) 505 kg
Color Anthracite Gray, Light Gray
Material Gypsum
Product Type Flammables Storage Cabinet