Learn More
Pyrvinium pamoate, MedChemExpress
Product Code. 16474197
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16474197
Brand: Medchem Express HYA0293
MedChemExpress Pyrvinium pamoate is an FDA-approved antihelmintic drug that inhibits WNT pathway signaling.
Pyrvinium pamoate is an FDA-approved antihelmintic drug that inhibits WNT pathway signaling.
Specifications
Chemical Name or Material Pyrvinium pamoate
Physical Form Solid
CAS 3546-41-6
Color Red
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C26H28N3.1/2C23H14O6
For Use With (Application) Cancer-programmed cell death
Synonym Pyrvinium embonate
Solubility Information DMSO : 17.86 mg/mL (31.02 mM; Need ultrasonic)
SMILES C[N+]1=C2C=CC(N(C)C)=CC2=CC=C1/C=C/C3=C(C)N(C4=CC=CC=C4)C(C)=C3.C[N+]5=C6C=CC(N(C)C)=CC6=CC=C5/C=C/C7=C(C)N(C8=CC=CC=C8)C(C)=C7.O=C(C9=C(O)C(CC%10=C%11C=CC=CC%11=CC(C([O-])=O)=C%10O)=C%12C=CC=CC%12=C9)[O-]
Molecular Weight (g/mol) 575.7
Formula Weight 575.7
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.