missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvate Dehydrogenase Kinase 1/PDK1 Antibody [CoraFluor™ 1], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2384CL1
This item is not returnable.
View return policy
Description
Pyruvate Dehydrogenase Kinase 1/PDK1 Polyclonal antibody specifically detects Pyruvate Dehydrogenase Kinase 1/PDK1 in Human,Mouse samples. It is validated for Western Blot,Immunoblot
Specifications
| Pyruvate Dehydrogenase Kinase 1/PDK1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoblot | |
| CoraFluor 1 | |
| EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, PDH kinase 1, PDK1, PDK-1, PKB kinase, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1, Pyruvate dehydrogenase lipoamide kinase isozyme 1, mitochondrial | |
| A synthetic peptide made to an internal portion of the human PDK1 protein sequence (between residues 300-400). [Swiss-Prot Q15118] | |
| 0.1 mL | |
| Autophagy, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction | |
| 5163 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction