Learn More
Pyrrole, Sigma-Aldrich™
Product Code. 15879490
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879490 25 mL 25mL
15836460 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15879490 Supplier Sigma Aldrich Supplier No. 8074920025
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-97-7
Specifications
Melting Point -24°C
Density 0.97 g/cm3 at 20°C
Boiling Point 130°C to 131°C/1013 hPa
MDL Number MFCD00005216
Quantity 25 mL
Synonym Pyrrole, Pyrrole
Molecular Weight (g/mol) 67.09
Grade Research