Learn More
Lonza™ Pyrogen-free Test Tubes
Product Code. 11610311
Change view
Click to view available options
Quantity:
50 Tubes
Unit Size:
Each
Product Code. Quantity unitSize
11610311 50 Tubes Each
1 options
Product Code. 11610311
Brand: Lonza™ N205
Only null left
Add to basket
Add to basket
Depyrogenated at extremely high temperatures well in excess of standard depyrogenation process conditions.
  • Pyrogen-free borosilicate glass test tubes without screw-caps.
  • Recommended for gel-clot testing.
  • Three packages with single vials front view.
  • Suitable for use when performing the bacterial endotoxins test (BET).
  • Certified to contain less than 0.005 EU/mL endotoxin.
  • All test tubes are made from USP Type 1 flint borosilicate glass.
TRUSTED_SUSTAINABILITY
Specifications
Description Type 1 flint borosilicate glass
Length (Metric) 75 mm
Material Borosilicate Glass
Quantity 50 Tubes
Diameter (Metric) 10 mm
Product Type Test Tube