Learn More
Pyriproxyfen, TRC
Specifications
Specifications
| Chemical Name or Material | Pyriproxyfen |
| CAS | 95737-68-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20 H19 N O3 |
| Quantity | 250 mg |
| Synonym | 2-[1-Methyl-2-(4-phenoxyphenoxy)ethoxy]pyridine, Admiral, Archer IGR, BCP 8702, Distance, Distance IGR, Esteem, Juventox, Juvinal, Knack, Lano Tape, Nemesis, NyGuard IGR, Nylar, OMS 3019, Pluto MC, Pyriproxyfen, S 31183, S 71639, S 9138, SK 591, Seize, Sumilarv, Tiger, Tiger (insecticide), Tiger 10EC |
| InChI Formula | InChI=1S/C20H19NO3/c1-16(23-20-9-5-6-14-21-20)15-22-17-10-12-19(13-11-17)24-18-7-3-2-4-8-18/h2-14,16H,15H2,1H3 |
| SMILES | CC(COc1ccc(Oc2ccccc2)cc1)Oc3ccccn3 |
| IUPAC Name | 2-[1-(4-phenoxyphenoxy)propan-2-yloxy]pyridine |
| Molecular Weight (g/mol) | 321.37 |
| Show More |