Learn More
Pyriproxyfen, MedChemExpress
Product Code. 16491138
Encompass_Preferred
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. 16491138
Brand: Medchem Express HYB2031
MedChemExpress Pyriproxyfen is a juvenile hormone analog, preventing larvae from developing into adulthood and thus rendering them unable to reproduce.
Pyriproxyfen is a juvenile hormone analog, preventing larvae from developing into adulthood and thus rendering them unable to reproduce.
Specifications
Chemical Name or Material Pyriproxyfen
CAS 95737-68-1
Color White
Purity Grade Notes Research
Molecular Formula C20H19NO3
Quantity 5 g
Synonym S-31183
Solubility Information DMSO : ≥ 100 mg/mL (311.17 mM)
SMILES CC(OC1=NC=CC=C1)COC2=CC=C(OC3=CC=CC=C3)C=C2
Molecular Weight (g/mol) 321.37
Formula Weight 321.37
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.