Learn More
Pyridine-2,6-dicarboxylic Acid, Sigma-Aldrich™
Specifications
Specifications
| CAS | 499-83-2 |
| Melting Point | 248°C to 255°C |
| Molecular Formula | 2,6-(HOOC)2C5H3N |
| MDL Number | MFCD00006299 |
| Quantity | 100 g |
| Synonym | Pyridine-2,6-dicarboxylic acid, Dipicolinic acid, Pyridine-2,6-dicarboxylic acid |
| Molecular Weight (g/mol) | 167.12 |
| Grade | Research |