missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,4-dicarbonitrile, 97%, Thermo Scientific™

Catalog Number 11319226
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
29181-50-8
C7H3N3
129.122
MFCD00014627
HLAGQMFURMNTLW-UHFFFAOYSA-N
2,4-dicyanopyridine, 2,4-pyridinedicarbonitrile, acmc-209wk1, 2 pound not4-pyridinedicarbonitrile, 2,4-pyridinedicarbonitrile 6ci,7ci,8ci,9ci
120144
pyridine-2,4-dicarbonitrile
C1=CN=C(C=C1C#N)C#N
This item is not returnable. View return policy
29181-50-8
C7H3N3
129.122
MFCD00014627
HLAGQMFURMNTLW-UHFFFAOYSA-N
2,4-dicyanopyridine, 2,4-pyridinedicarbonitrile, acmc-209wk1, 2 pound not4-pyridinedicarbonitrile, 2,4-pyridinedicarbonitrile 6ci,7ci,8ci,9ci
120144
pyridine-2,4-dicarbonitrile
C1=CN=C(C=C1C#N)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 87°C to 92°C
Quantity 250 mg
Beilstein 115936
Formula Weight 129.12
Percent Purity 97%
Chemical Name or Material Pyridine-2,4-dicarbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'