Learn More
Pyrex™ T-bore Plain arms 3-way Stopcock with PTFE Key
Product Code. p-8341000
Click to view available options
Diameter (Metric):
14.5 mm
18.8 mm
Plug Bore:
2.5 mm
4 mm
Unit Size:
Pack of 5
Product Code. 12098244
Brand: Pyrex SST304
Only null left
Add to basket
Add to basket
3-way Stopcock with PTFE Key
TRUSTED_SUSTAINABILITY
Specifications
Type Plain Arms Stopcock
Plug Bore 4 mm
Diameter (Metric) 18.8 mm
Includes PTFE Key