Learn More
Pyrex™ T-bore Plain arms 3-way Stopcock with PTFE Key
Product Code. 12098244
Change view
Click to view available options
Diameter (Metric):
14.5 mm
18.8 mm
Plug Bore:
2.5 mm
4 mm
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Plug Bore Unit Size
12098244 18.8 mm 4 mm Pack of 5
10775701 14.5 mm 2.5 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12098244 Supplier Pyrex Supplier No. SST304
  • ONLINE EXCLUSIVE PRICE
    6069.00 NOK
    5516.22 NOK / Pack of 5
    Save 552.78 NOK
    9% Off
Estimated Shipment: 17-11-2026
3-way Stopcock with PTFE Key
Specifications
Type Plain Arms Stopcock
Plug Bore 4 mm
Diameter (Metric) 18.8 mm
Includes PTFE Key