Learn More
Pyrex™ Screwcap Spare Inserts
For use with Black SVL™ PBT screwcaps
Brand: Pyrex 4513/22M
Additional Details : Weight : 0.01100kg
Description
- PTFE faced sealing disc for chemical resistance
- Autoclavable up to 121°C
Specifications
Screw Cap Insert | |
22 mm | |
PTFE, Rubber |