Learn More
Pyrex™ Screwcap Spare Inserts
For use with Black SVL™ PBT screwcaps
Brand:  Pyrex 4513/22M
441.75 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 10788015
  • 465.00 NOK / Pack of 20
Estimated Shipment: 08-08-2024
Description
  • PTFE faced sealing disc for chemical resistance
  • Autoclavable up to 121°C
Specifications
Specifications
Screw Cap Insert
22 mm
PTFE, Rubber