Learn More
Pyrex™ Roux Culture Off-Set Neck Flasks
Product Code. p-8338041
Change view
Click to view available options
Capacity (Metric):
600 mL
1200 mL
Unit Size:
Pack of 10
Product Code. Capacity (Metric) unitSize
12574257 1200 mL Pack of 10
13411859 600 mL Pack of 10
2 options
Product Code. 12574257
Brand: Pyrex 1350/06M
Ideal for cell culture
  • Manufactured from chemically inert Pyrex glass for a contamination-free environment
  • Withstands repeated autoclaving /li>
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1200 mL
Material Borosilicate Glass
For Use With (Application) Ideal for culture work
Neck Style Off-set
No. per Pack 10