Learn More
Corning™ PYREX™ Round-Bottom Glass Centrifuge Tubes
Product Code. 12704868
Change view
Click to view available options
Length (Metric):
122 mm
Closure Included:
Yes
Closure Material:
Phenolic
Unit Size:
Pack of 12
Product Code. Length (Metric) Closure Included Closure Material unitSize
12704868 122 mm Yes Phenolic Pack of 12
1 options
Product Code. 12704868
Brand: Corning™ 842250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Without graduations
These PYREX™ round bottom, ungraduated centrifuge tubes have a pourout spout for easy decanting.
TRUSTED_SUSTAINABILITY
Specifications
Closure Material Phenolic
Capacity (Metric) 50 mL
Disposable Reusable
Material Borosilicate Glass
For Use With Useful for the handling of sputum specimens, digestion, shaking, neutralizing and centrifuging
Liner Material Rubber
No. per Case 12
No. per Pack 12
Thread Size 24-410
Sterility Non-sterile
Shape Cylindrical
Length (Metric) 122 mm
Closure Included Yes
Diameter (Metric) 29 mm
Product Type Centrifuge Tube