Learn More
Corning™ PYREX™ Polypropylene Flask Closures
Product Code. 15404633 Shop All Corning Products
Change view
Click to view available options
Diameter (Metric) Outer Neck:
25 mm
3.8 cm
Closure Color:
Natural
Orange
Unit Size:
Each
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Neck Closure Color unitSize
15404633 3.8 cm Orange Each
17213494 3.8 cm Natural Pack of 100
17203494 25 mm Natural Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15404633 Supplier Corning™ Supplier No. 2679638
  • ONLINE EXCLUSIVE PRICE
    2365.00 NOK
    2208.68 NOK / Pack of 1
    Save 156.32 NOK
    6% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
Designed for culture tubes and flasks with long rimless necks
These Corning polypropylene closures are designed for culture tubes and flasks with long rimless necks. Closure offers two positions: open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination. Closures are autoclavable at 121°C.
Closure ending in -25 fits Cat. No. 4442-125.
Closure ending with -38 fits Cat. Nos. 4442-250, 4442-500, 4442-1 L, and 4442-2L.
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Orange
Closure Material Polypropylene
Color Orange
Diameter (Metric) Outer Neck 3.8 cm
For Use With (Equipment) Culture tubes and flasks with long ri mLess necks
No. per Case 100/Cs.
No. per Pack 50/Pk.
Product Type Cap, Flask
Autoclavable Autoclavable
Disposable Reusable