Learn More
Pyrex™ Labware Polyethylene Reservoir Bottle
Feed liquids into a Sofnol burette with easy this filling squeeze bottle. The Pyrex™ labware polyethylene reservoir bottle is a replacement part for the Sofnol, auto zero, picnch clip, class B burette
Brand: Pyrex 3299/04
124.45 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.08366kg
Description
- Polyethylene
- Squeeze bottle
- For use with Sofnol burette
- Squeeze the bottle to fill the burette
- Release pressure to allow excess liquid to drain back into bottle and set the burrette to zero
- Used for liquid testing
Specifications
500 mL | |
Sofnol Burette | |
Polyethylene | |
Reservoir Bottle |