Learn More
Pyrex™ Hexagonal Head Solid Glass Stopper
Description
- Robust, solid glass construction
- Hexagonal head to provide a good grip
Specifications
Specifications
| For Use With (Equipment) | Pyrex bottles and aspirators |
| Standard Taper Joint | 19/26 |
| Material | Glass |
| Product Type | Hexagonal Head Stopper |