Learn More
PYREX™ Heavy-Duty Griffin Beakers
GREENER_CHOICE
Product Code. p-33772
Click to view available options
Capacity (Metric):
150 mL
250 mL
400 mL
600 mL
1 L
2000 mL
3000 mL
4000 mL
5000 mL
Unit Size:
Each
Pack of 12
Pack of 4
Pack of 6
Product Code. 10079219
Brand: Pyrex 1003150
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use under harsh conditions such as mechanized washing
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes.
Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot.
Compliance
Designed to ASTM™ E 960 requirements.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 5.07 oz.
Diameter (English) Outer 2.24 in.
Diameter (Metric) Outer 5.7 cm
Capacity (Metric) 150 mL
Material Borosilicate Glass
For Use With (Application) Provides The Best Mechanical Strength Under Harsh Conditions Such as Mechanized Washings
Graduation Range 20 to 140 mL
Height (English) 3.38 in.
Height (Metric) 8.6 cm
Increments 20 mL
Label Type Extra Large Marking
Quantity 12 Pk.
Type Beaker
Disposable Reusable
Graduated Yes
Certifications/Compliance ASTM E960
Scale Graduated Metric Scale
Style Griffin