Learn More
PYREX™ Heavy-Duty Griffin Beakers
Product Code. 10819610
Change view
Click to view available options
Capacity (Metric):
150 mL
250 mL
400 mL
600 mL
1 L
2000 mL
3000 mL
4000 mL
5000 mL
Unit Size:
Each
Pack of 12
Pack of 4
Pack of 6
Product Code. Capacity (Metric) unitSize
10819610 400 mL Each
10079219 150 mL Each
12549995 150 mL Pack of 12
12559995 250 mL Pack of 12
10734192 250 mL Each
12569995 400 mL Pack of 12
12671766 600 mL Pack of 6
10680941 1 L Each
12509975 1 L Pack of 6
11911516 2000 mL Pack of 4
10566370 2000 mL Each
10343010 3000 mL Each
10337879 4000 mL Each
10829610 5000 mL Each
14 options
Product Code. 10819610
Brand: Pyrex 1003400
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use under harsh conditions such as mechanized washing
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes.
Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot.
Compliance
Designed to ASTM™ E 960 requirements.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 13.52 oz.
Diameter (English) Outer 3.22 in.
Diameter (Metric) Outer 8.2 cm
Capacity (Metric) 400 mL
Material Borosilicate Glass
For Use With (Application) Provides The Best Mechanical Strength Under Harsh Conditions Such as Mechanized Washings
Graduation Range 25 to 325 mL
Height (English) 4.33 in.
Height (Metric) 11 cm
Increments 25 mL
Label Type Extra Large Marking
Quantity 12 Pk.
Type Beaker
Disposable Reusable
Graduated Yes
Certifications/Compliance ASTM E960
Scale Dual Graduated Metric Scale
Style Griffin