Learn More
PYREX™ Heavy-Duty Griffin Beakers
Description
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes.
Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot.
Compliance
Designed to ASTM™ E 960 requirements.
Specifications
Specifications
| Capacity (English) | 5.07 oz. |
| Diameter (English) Outer | 2.24 in. |
| Diameter (Metric) Outer | 5.7 cm |
| Capacity (Metric) | 150 mL |
| Material | Borosilicate Glass |
| For Use With (Application) | Provides The Best Mechanical Strength Under Harsh Conditions Such as Mechanized Washings |
| Graduation Range | 20 to 140 mL |
| Height (English) | 3.38 in. |
| Height (Metric) | 8.6 cm |
| Increments | 20 mL |
| Show More |