Learn More
Pyrex™ Gas Distribution Tube
Product Code. 10217634
Change view
Click to view available options
Description:
Porosity: 0
Porosity: 1
Porosity: 2
Porosity: 3
Unit Size:
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10217634 Porosity: 0 Pack of 2
15608166 Porosity: 2 Pack of 2
12663232 Porosity: 1 Pack of 2
12547866 Porosity: 3 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10217634 Supplier Pyrex Supplier No. 3830/00M
  • ONLINE EXCLUSIVE PRICE
    1092.00 NOK
    899.56 NOK / Pack of 2
    Save 192.44 NOK
    17% Off
Estimated Shipment: 25-01-2027
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
  • Ideal for the dispersion of gas in liquids
  • Borosilicate glass tube with sintered thimble
  • Choice of thimble porosities to achieve optimum gas bubble size
Specifications
Description Porosity: 0
Material Borosilicate Glass (3.3)
Overall Length (Metric) 300 mm
For Use With (Application) For the dispersion of gas in liquids
Includes Sintered thimble