Learn More
Pyrex™ Funnel Repair Stopcocks with PTFE Key
Product Code. 13491899
Change view
Click to view available options
Plug Bore:
1.5 mm Key Bore
2.5 mm Key Bore
4 mm Key Bore
6 mm Key Bore
Unit Size:
Pack of 5
Product Code. Plug Bore unitSize
13491899 6 mm Key Bore Pack of 5
13481899 4 mm Key Bore Pack of 5
12451021 2.5 mm Key Bore Pack of 5
13471899 1.5 mm Key Bore Pack of 5
4 options
Product Code. 13491899
Brand: Pyrex SST706
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Contains one flared end for connection to the base of the funnel
  • Contains one flared end for connection to the base of the funnel and 100mm long stem with angled tip
  • Straight bore
TRUSTED_SUSTAINABILITY
Specifications
Type Funnel Repair
Plug Bore 6 mm Key Bore
Diameter (Metric) 18.8 mm
Material Glass
Includes PTFE key