Learn More
Pyrex™ ETFE Pouring Rings
Product Code. p-8002583
Click to view available options
Thread Size (Metric):
32 mm
45 mm
Unit Size:
Pack of 10
Product Code. 11398044
Brand: Pyrex™ 4536/32D
Only null left
Add to basket
Add to basket
Prevent drips while pouring. Pyrex™ ETFE Pouring Rings work well with high temperature applications.
  • For use with 1516, 1517, 1518, 1519 and 1520 series bottles
  • Ideal for applications involving high temperatures
  • Red colored ethylene-tetrafluoroethylene pouring ring
  • Withstands hot air sterilisation up to 180°C
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Material ETFE
Quantity 10/PK.
Sterility Dry Heat Sterilized
Thread Size (Metric) 32 mm
For Use With (Equipment) Media-lab 1516, 1517, 1518, 1519 and 1520 series bottles