Learn More
Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes
Product Code. 13033073 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
4 mL
6 mL
10 mL
11 mL
15 mL
19 mL
23 mL
28.5 mL
36 mL
Length (Metric):
75 mm
85 mm
100 mm
125 mm
150 mm
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
Product Code. Capacity (Metric) Length (Metric) unitSize
13033073 6 mL 75 mm Pack of 250
10212563 19 mL 125 mm Pack of 1000
13073073 19 mL 125 mm Pack of 250
13023073 4 mL 75 mm Pack of 250
10672234 4 mL 75 mm Pack of 1000
13043073 10 mL 100 mm Pack of 250
10672424 6 mL 75 mm Pack of 1000
13053073 11 mL 85 mm Pack of 250
10692424 11 mL 85 mm Pack of 1000
12079257 36 mL 150 mm Pack of 250
10692614 36 mL 150 mm Pack of 500
13083073 23 mL 150 mm Pack of 250
10018522 23 mL 150 mm Pack of 1000
13063073 15 mL 100 mm Pack of 250
10008522 15 mL 100 mm Pack of 1000
11992288 28.5 mL 150 mm Pack of 250
10735763 28.5 mL 150 mm Pack of 500
17 options
Product Code. 13033073
Brand: Corning™ 9944512
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed. Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 6 mL
Length (Metric) 75 mm
Material Glass
Disposable Single-use
For Use With For tissue culture work and general bacteriological use
Label Type None
Packaging Bulk
Rimmed No
Sterility Non-sterile
Graduated No
Diameter (Metric) 12 mm
Product Type Culture Tube