Learn More
Pyrex™ Cylindrical Centrifuge Tubes
Centrifuge in sturdy glass and chemically resistant tubes. Pyrex™ Cylindrical Centrifuge Tubes come with a round base.
Brand: Pyrex 1660/06M
2479.50 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 1.06000kg
Description
- Cylindrical type, round base
- Uniform glass distribution in wall and base for improved strength
- Manufactured from chemically resistant Pyrex™ glass
- Can be centrifuged at RCF values up to 3, 000
Specifications
25 mL | |
40 | |
90 mm | |
3000 x g |
Borosilicate Glass | |
Cylindrical | |
24 mm |