Learn More
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Description
- Complies with ISO 4788
- Calibrated to Class A tolerances
- Individually tested and supplied with certificate showing exact volume contained at 5 points on the scale
- Conformity H certified
- Printed with individual serial number for batch certification
- Manufactured from chemically resistant Pyrex borosilicate glass with hexagonal glass
- Graduations in white enamel for improved visibility with dark colored solutions
- Supplied with polyethylene stopper
Specifications
Specifications
| Base Type | Hexagonal |
| Type | Graduated Cylinder |
| Material | Glass |
| Certifications/Compliance | ISO 4788 |
| Class | A |
| Capacity (Metric) | 250 mL |
| Height (Metric) | 367 mm |
| Includes | Polyethylene Stopper |
| Stopper Number | 29/32 |
| Stopper Material | Polyethylene |