Learn More
Pyrex™ Wide Neck Round Bottom Boiling Flask
Product Code. p-8002572
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
4 L
6 L
Unit Size:
Each
Pack of 10
Pack of 30
Pack of 5
Product Code. 12539105
Brand: Pyrex 1120/08D
Only null left
Add to basket
Add to basket
Made of borosilicate glass
  • Complies with ISO 24450 and DIN 12353
  • With wide neck to facilitate addition of solid samples etc
  • Resistant to chemical corrosion and thermal shock
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 24450, DIN 12353
Capacity (Metric) 250 mL
Material Borosilicate Glass
Height (Metric) 145 mm
Neck Style Wide
Bottom Shape Round
Diameter (Metric) Outer 85 mm
Closure Size 45