Learn More
Pyrex™ Borosilicate Glass Wide Neck Flat Bottom Boiling Flask Product Code.: 10427230
Estimated Shipment: 05-01-2026
Only null left
Add to basket
Pyrex™ Borosilicate Glass Wide Neck Flat Bottom Boiling Flask
Product Code. p-8401023
250 mL, Pack of 10
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 10
Pack of 30
Product Code. 10427230
Brand: Pyrex 1080/08D
Only null left
Estimated Shipment: 05-01-2026
Add to basket
Only null left
Add to basket
Manufactured from Pyrex™ glass for high resistance to thermal shock
  • Complies with ISO 24450
  • With wide neck to facilitate easy addition of solid samples etc
  • Flat base for standing on benches and hotplates etc
  • Manufactured from Pyrex™ glass for high resistance to thermal shock
  • Excellent chemical durability for contamination free performance
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 24450
Capacity (Metric) 250 mL
Material Borosilicate Glass
Height (Metric) 140 mm
Product Content Correction
Product Title
Pyrex™ Borosilicate Glass Wide Neck Flat Bottom Boiling Flask >