Learn More
Pyrex™ Borosilicate Glass Wide Neck Flat Bottom Boiling Flask
Product Code. 10427230
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
1000 mL
2000 mL
Packungsgröße:
Pack of 10
Pack of 30
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10427230 250 mL Pack of 10
12539095 500 mL Pack of 10
12549095 1000 mL Pack of 10
12423431 2000 mL Pack of 10
15547660 50 mL Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10427230 Supplier Pyrex Supplier No. 1080/08D
Only null left
Add to basket
Add to basket
Manufactured from Pyrex™ glass for high resistance to thermal shock
  • Complies with ISO 24450
  • With wide neck to facilitate easy addition of solid samples etc
  • Flat base for standing on benches and hotplates etc
  • Manufactured from Pyrex™ glass for high resistance to thermal shock
  • Excellent chemical durability for contamination free performance
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 24450
Capacity (Metric) 250 mL
Material Borosilicate Glass
Height (Metric) 140 mm