Learn More
Pyrex™ Borosilicate Glass Watch Glasses
Product Code. 11303454
Change view
Click to view available options
Diameter (Metric):
40 mm
50 mm
60 mm
80 mm
100 mm
125 mm
150 mm
200 mm
250 mm
Unit Size:
Each
Pack of 10
Product Code. Diameter (Metric) unitSize
11303454 100 mm Each
10468370 100 mm Pack of 10
10651651 125 mm Pack of 10
11313454 125 mm Each
10315100 150 mm Pack of 10
11323454 150 mm Each
10101780 250 mm Pack of 10
11367824 250 mm Each
11373444 50 mm Each
10706071 50 mm Pack of 10
11363444 40 mm Each
10681101 40 mm Pack of 10
10706261 60 mm Pack of 10
11383444 60 mm Each
11926288 80 mm Pack of 10
11393444 80 mm Each
13481829 200 mm Pack of 10
17 options
Product Code. 11303454
Brand: Pyrex 1060/14D
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Watch Glasses are used for evaporation work and covering heated beakers. Fire-polished edges reduce chipping and breakage.
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex™ borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
TRUSTED_SUSTAINABILITY
Specifications
Bottom Shape Flat
Material Borosilicate Glass
Shape Round
Type Watch Glass
For Use With (Application) Evaporation Work and Covering Heated Beakers
Interior Finish Polished
Diameter (Metric) 100 mm