Learn More
Pyrex™ Borosilicate Glass Vacuum Desiccator Cover
Product Code. 11985869
Change view
Click to view available options
For Use With (Equipment):
Vacuum desiccators
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11985869 Vacuum desiccators Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11985869 Supplier Pyrex Supplier No. 1594/04D
  • ONLINE EXCLUSIVE PRICE
    3375.00 NOK
    2779.81 NOK / Pack of 1
    Save 595.19 NOK
    17% Off
Estimated Shipment: 30-03-2027
Designed for use under vacuum.
Designed for use under vacuum.
  • Designed for use under vacuum
  • Sturdy construction in borosilicate glass
  • Ground glass flanges
  • Cover has a 24/29 socket to accept desiccator stopcocks
  • Accommodates 200mm diameter plates
  • Plates are positioned on an internal ledge within the base
  • Accessory metal perforated plate and Rotaflo™ glass/PTFE stopcock available
Specifications
Diameter (Metric) Outer 220 mm
Material Borosilicate Glass
For Use With (Equipment) Vacuum desiccators
Product Type Vacuum Desiccator Cover