Learn More
Pyrex™ Borosilicate Glass Plain Neck Kjeldahl Flask
Product Code. 11313624
Change view
Click to view available options
Capacity (Metric):
100 mL
300 mL
500 mL
800 mL
Diameter (Metric) Outer:
101 mm
115 mm
81 mm
Bulb: 58 mm
Unit Size:
Pack of 10
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer unitSize
11313624 800 mL 115 mm Pack of 10
11303624 500 mL 101 mm Pack of 10
11383614 100 mL Bulb: 58 mm Pack of 10
15547680 300 mL 81 mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11313624 Supplier Pyrex Supplier No. 1200/16M
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
Suitable for the determination of nitrogen
  • Withstands high temperatures
  • Resistant to chemical corrosion
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner Neck 30 mm
Diameter (Metric) Outer Neck 34 mm
Capacity (Metric) 800 mL
Material Borosilicate Glass
Height (Metric) 350 mm
Neck Style Plain
Diameter (Metric) Outer 115 mm