Learn More
Pyrex™ Borosilicate Glass Non-vacuum Desiccators
Product Code. 12466947
Change view
Click to view available options
Height (Metric):
85 mm
120 mm
Diameter (Metric) Outer:
170 mm
220 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Diameter (Metric) Outer unitSize
12466947 85 mm 170 mm Each
12476947 120 mm 220 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12466947 Supplier Pyrex Supplier No. 1590/02D
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
Simple design for drying under atmospheric pressure.
  • Robust desiccator base and cover in borosilicate glass
  • Ground glass flanges
  • Plates are positioned on an internal ledge within the base
  • Choice of 2 sizes - to accommodate 150mm or 200mm diameter plates
  • Accessory metal perforated plate available
TRUSTED_SUSTAINABILITY
Specifications
Product Type Nonvacuum Desiccator
Includes Complete desiccator
Size Plate: 150 mm
Height (Metric) 85 mm
Diameter (Metric) Outer 170 mm