Learn More
Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes
Product Code. 11317904
Change view
Click to view available options
Diameter (Metric):
10 mm
12 mm
16 mm
18 mm
24 mm
Length (Metric):
75 mm
100 mm
125 mm
150 mm
180 mm
200 mm
Capacity (Metric):
4 mL
6 mL
8 mL
14 mL
18 mL
22 mL
28 mL
34 mL
55 mL
73 mL
Unit Size:
Each
Pack of 100
Pack of 50
Product Code. Diameter (Metric) Length (Metric) Capacity (Metric) unitSize
11317904 18 mm 180 mm 34 mL Each
12549485 24 mm 150 mm 55 mL Pack of 100
11337904 24 mm 150 mm 55 mL Each
12539475 18 mm 150 mm 28 mL Pack of 100
12559475 24 mm 200 mm 73 mL Pack of 50
11347904 24 mm 200 mm 73 mL Each
12653192 18 mm 180 mm 34 mL Pack of 100
12599465 16 mm 150 mm 22 mL Pack of 100
12529485 12 mm 75 mm 6 mL Pack of 100
12519465 10 mm 75 mm 4 mL Pack of 100
12529465 12 mm 100 mm 8 mL Pack of 100
12579465 16 mm 100 mm 14 mL Pack of 100
12589465 16 mm 125 mm 18 mL Pack of 100
13 options
Product Code. 11317904
Brand: Pyrex 1620/15M
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Medium walled test tubes with rimmed edgeManufactured from Pyrex™ borosilicate glass to give superior performance over neutral borosilicate and soda lime glass and excellent chemical resistance. Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes are ideal for microbiology culture work.
Manufactured from Pyrex™ borosilicate glass to give superior performance over neutral borosilicate and soda lime glass and excellent chemical resistance. Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes are ideal for microbiology culture work.
  • Rimmed medium walls: constructed for optimum performance
  • Robust construction: allows use at high temperatures up to 500deg.C
  • Conforms to: ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 4142
Capacity (Metric) 34 mL
Length (Metric) 180 mm
Material Borosilicate Glass
Quantity 100 Pack
Rimmed Yes
Diameter (Metric) 18 mm