Learn More
Pyrex™ Quickfit™ Borosilicate Glass Florentine Flask
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Standard Taper Joint:
29/32
Height (Metric):
107 mm
151 mm
160 mm
198 mm
Unit Size:
Each
Description
Remove solid residues with ease, using this robust, pear-shaped flask. Quickfit™ Borosilicate Glass Florentine Flask features a large surface area for rapid evaporation.
- Ideal for use with rotary evaporators
- Large surface area for rapid evaporation
- Robust heavy wall construction for added strength
- Shape allows easy removal of solid residues
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 151 mm |
| Standard Taper Joint | 29/32 |